Learn More
CD8 Mouse anti-Human, PE, Clone: SK1, Proteintech
Mouse Monoclonal Antibody
Merk: Proteintech PE-65146-100TESTS
This item is not returnable.
View return policy
Beschrijving
The CD8 antibody from Proteintech is a mouse monoclonal antibody generated with n/a of human CD8. This antibody recognizes human antigen. The CD8 antibody has been validated for the following applications: FC analysis. This antibody is pe.
Specificatie
CD8 | |
Monoclonal | |
5 μL (0.125 μg)/Test | |
Mouse | |
Primary | |
Human |
Flow Cytometry | |
SK1 | |
PE | |
100 Tests | |
925 | |
IgG1 |