Learn More
CD3ε Hamster anti-Mouse, FITC, Clone: 145-2C11, Proteintech
Hamster Monoclonal Antibody
Merk: Proteintech FITC-65060-100UG
This item is not returnable.
View return policy
Beschrijving
The CD3ε antibody from Proteintech is a hamster monoclonal antibody generated with h-2kb-specific murine cytotoxic t-lymphocyte (ctl) clone bm10-37 of mouse CD3ε. This antibody recognizes mouse antigen. The CD3ε antibody has been validated for the following applications: FC analysis. This antibody is fitc.
Specificatie
CD3ε | |
Monoclonal | |
0.1 mg/mL | |
Hamster | |
Primary | |
Mouse |
Flow Cytometry | |
145-2C11 | |
FITC | |
100 μg | |
12501 | |
IgG |