Learn More
CD3ε Hamster anti-Mouse, FITC, Clone: 145-2C11, Proteintech
Hamster Monoclonal Antibody
Merk:  Proteintech FITC-65060-100UG
Produktcode. 16861176
  • 37.00 /
Explore more special offers
Beschrijving
Beschrijving
The CD3ε antibody from Proteintech is a hamster monoclonal antibody generated with h-2kb-specific murine cytotoxic t-lymphocyte (ctl) clone bm10-37 of mouse CD3ε. This antibody recognizes mouse antigen. The CD3ε antibody has been validated for the following applications: FC analysis. This antibody is fitc.
Specificatie
Specificatie
CD3ε
Monoclonal
0.1 mg/mL
Hamster
Primary
Mouse
Flow Cytometry
145-2C11
FITC
100 μg
12501
IgG
Product Suggestions
Product Suggestions
Video's
SDS
Documentatie
Documentatie
Certificates
Speciale aanbiedingen
Speciale aanbiedingen