Learn More
CD19 Mouse anti-Human, APC, Clone: HIB19, Proteintech
Mouse Monoclonal Antibody
Merk: Proteintech APC-65110-100TESTS
This item is not returnable.
View return policy
Beschrijving
The CD19 antibody from Proteintech is a mouse monoclonal antibody generated with n/a of human CD19. This antibody recognizes human antigen. The CD19 antibody has been validated for the following applications: FC analysis. This antibody is apc.
Specificatie
CD19 | |
Monoclonal | |
5 μL (0.125 μg)/Test | |
Mouse | |
Primary | |
Human |
Flow Cytometry | |
HIB19 | |
APC | |
100 Tests | |
930 | |
IgG1 |