Learn More
CD161 Mouse anti-Human, CoraLite 488, Clone: HP-3G10, Proteintech
Mouse Monoclonal Antibody
Merk: Proteintech CL488-65115-100TESTS
This item is not returnable.
View return policy
Beschrijving
The CD161 antibody from Proteintech is a mouse monoclonal antibody generated with human nk cells of human CD161. This antibody recognizes human antigen. The CD161 antibody has been validated for the following applications: FC analysis. This antibody is coralite™488.
Specificatie
CD161 | |
Monoclonal | |
5 μL/Test | |
Mouse | |
Primary | |
Human |
Flow Cytometry | |
HP-3G10 | |
CoraLite 488 | |
100 Tests | |
3820 | |
IgG1 |