Learn More
CCS Mouse, anti-Human, CoraLite Plus 488, Clone: 3F5E5, Proteintech
Mouse Monoclonal Antibody
Merk: Proteintech CL488-68341-20UL
This item is not returnable.
View return policy
Beschrijving
The CCS antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CCS. This antibody recognizes human, mouse, rat antigen. The CCS antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specificatie
CCS | |
3F5E5 | |
CoraLite Plus 488 | |
Mouse | |
Primary | |
Human, Mouse, Rat | |
155 kDa | |
IgG2a |
Immunofluorescence | |
1000 μg/mL | |
CCS | |
20 μL | |
9973 | |
Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
Liquid |