Learn More
CCS Mouse, anti-Human, CoraLite Plus 488, Clone: 3F5E5, Proteintech
Mouse Monoclonal Antibody
Merk:  Proteintech CL488-68341-100UL
 Bekijk meerdere types van dit product via de
Produktcode. 18077415
  • 450.00 /
Explore more special offers
Beschrijving
Beschrijving
The CCS antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CCS. This antibody recognizes human, mouse, rat antigen. The CCS antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specificatie
Specificatie
CCS
3F5E5
CoraLite Plus 488
Mouse
Primary
Human, Mouse, Rat
155 kDa
IgG2a
Immunofluorescence
1000 μg/mL
CCS
100 μL
9973
Store at -20°C. Avoid exposure to light. Stable for one year after shipment.
Liquid
Product Suggestions
Product Suggestions
Video's
SDS
Documentatie
Documentatie
Certificates
Speciale aanbiedingen
Speciale aanbiedingen