Learn More
CAP2 Mouse anti-Human, Unconjugated, Clone: 1G7D2, Proteintech
Mouse Monoclonal Antibody
Merk: Proteintech 67412-1-IG-20UL
This item is not returnable.
View return policy
Beschrijving
The CAP2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CAP2. This antibody recognizes human antigen. The CAP2 antibody has been validated for the following applications: WB, ELISA analysis.
Specificatie
CAP2 | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human |
Western Blot, ELISA | |
1G7D2 | |
Unconjugated | |
20 μL | |
10486 | |
IgG2a |