Learn More
Calumenin Mouse anti-Human, Unconjugated, Clone: 2D8B8, Proteintech
Mouse Monoclonal Antibody
Merk:  Proteintech 67585-1-IG-150UL
 Bekijk meerdere types van dit product via de
Produktcode. 16850296
  • 410.00 /
Explore more special offers
Beschrijving
Beschrijving
The Calumenin antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human Calumenin. This antibody recognizes human, mouse, rat antigen. The Calumenin antibody has been validated for the following applications: IF, WB, ELISA analysis.
Specificatie
Specificatie
Calumenin
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Rat, Mouse
Western Blot, Immunofluorescence, ELISA
2D8B8
Unconjugated
150 μL
813
IgG1
Product Suggestions
Product Suggestions
Video's
SDS
Documentatie
Documentatie
Certificates
Speciale aanbiedingen
Speciale aanbiedingen