Learn More
Telstar™ Mini-V/PCR
Delivers an ultra clean laminar airflow over the working bench to protect manipulated products from dust, from environment contamination and from cross contamination in the working area. Telstar™ Mini-V/PCR includes a built–in UV light with timer is particularly suitable for PCR process to denaturate oligonucleotide contaminants.
Brand: Telstar™ 11215
Description
Delivers an ultra clean laminar airflow over the working bench to protect manipulated products from dust, from environment contamination and from cross contamination in the working area. Ideal solution for the handling of non-hazardous biological samples. A built–in UV light with timer is particularly suitable for PCR process to denaturate oligonucleotide contaminants. Bench top cabinet with non-recirculated vertical laminar flow.
- Compact dimensions compatible with all laboratory doors and dimensions.
- Epoxy powder coated finish zinctec steel cabinet.
- AISI 304 stainless steel work bench.
- H14 HEPA filter with 99,999 % efficiency for particles >0.3 micron easily accessible for replacement.
- Prefilter 85% ASHRAE, in the inlet side.
- Class 100 work station according to Fed. Std. 209 and BS 5295.
Specifications
Epoxy Powder | |
Class 100 work station according to Fed. Std. 209 and BS 5295 | |
500 x 630 x 570 mm | |
50 Hz | |
Electronic control system with LED display, fluorescent light, built-in UV light with timer, electrical socket in working area, stainless steel bench. | |
230 V | |
670 mm |
Epoxy Powder-coated Zinctec Steel, Stainless Steel Workbench | |
520 mm | |
1.125 m | |
>750 lx | |
56 dBa | |
70 kg |