Learn More
C1QBP Rabbit anti-Human, Unconjugated, Clone: 3K20, Proteintech
Rabbit Recombinant Antibody
Merk:  Proteintech 80490-1-RR-20UL
 Bekijk meerdere types van dit product via de
Produktcode. 17551506
  • 149.00 /
Beschrijving
Beschrijving
The C1QBP antibody from Proteintech is a rabbit recombinant antibody to a recombinant protein of human C1QBP. This antibody recognizes human, mouse, rat antigen. The C1QBP antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specificatie
Specificatie
C1QBP
3K20
Rabbit
Primary
Human, Mouse, Rat
Western Blot, Immunohistochemistry, Immunofluorescence
Unconjugated
20 μL
708
IgG
Product Suggestions
Product Suggestions
Video's
SDS
Documentatie
Documentatie