Learn More
BORIS Mouse anti-Human, CoraLite 594, Clone: 2B12E2, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL594-60079-100UL
Description
The BORIS antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human BORIS. This antibody recognizes human, mouse antigen. The BORIS antibody has been validated for the following applications: IF analysis. This antibody is coralite™594.
Specifications
BORIS | |
Monoclonal | |
2000 μg/mL | |
Mouse | |
Primary | |
Human, Mouse |
Immunofluorescence | |
2B12E2 | |
CoraLite 594 | |
100 μL | |
140690 | |
IgG1 |