Learn More
Ohaus Balance Compass CX221, 220g x 0.1g, External Calibration
Merk:  Ohaus 30428208
70.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Balance Compass CX221, 220g x 0.1g, External Calibration
Produktcode. 16203292
  • 74.75 / 1 eenheid