Learn More
B4GALT2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Merk: Proteintech 20330-1-AP-150UL
This item is not returnable.
View return policy
Beschrijving
B4GALT2 is one of type II membrane-bound glycoproteins that appear to have exclusive specificity for the donor substrate UDP-galactose; all transfer galactose in a beta1,4 linkage to similar acceptor sugars: GlcNAc, Glc, and Xyl. Each beta4GalT has a distinct function in the biosynthesis of different glycoconjugates and saccharide structures. As type II membrane proteins, they have an N-terminal hydrophobic signal sequence that directs the protein to the Golgi apparatus and which then remains uncleaved to function as a transmembrane anchor. By sequence similarity, the beta4GalTs form four groups: beta4GalT1 and beta4GalT2, beta4GalT3 and beta4GalT4, beta4GalT5 and beta4GalT6, and beta4GalT7. The enzyme synthesizes N-acetyllactosamine in glycolipids and glycoproteins. Its substrate specificity is affected by alpha-lactalbumin but it is not expressed in lactating mammary tissue.Specificatie
B4GALT2 | |
Polyclonal | |
Unconjugated | |
B4GALT2 | |
B4Gal T2, B4Gal T3, B4GALT2, Beta 1,4 GalTase 2, beta4Gal T2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
313536, 53418, 8704 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.28 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O60909, Q9Z2Y2 | |
B4galt2 | |
B4GALT2 Fusion Protein Ag14046 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |