Learn More
B3GNT5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Merk:  Proteintech 20422-1-AP-20UL
 Bekijk meerdere types van dit product via de
Produktcode. 16819964
  • 145.00 /
Beschrijving
Beschrijving
B3GNT5 is a member of the beta-1, 3-N-acetylglucosaminyltransferase family. This enzyme is a type II membrane protein. It exhibits strong activity to transfer GlcNAc to glycolipid substrates and is identified as the most likely candidate for lactotriaosylceramide synthase. This enzyme is essential for the expression of Lewis X epitopes on glycolipids.
Specificatie
Specificatie
B3GNT5
Polyclonal
Unconjugated
B3GNT5
B3GN T5, B3GNT5, Beta 1,3 Gn T5, beta3Gn T5, BGnT 5, Lactotriaosylceramide synthase, Lc(3)Cer synthase, Lc3 synthase
Rabbit
Antigen Affinity Chromatography
RUO
108105, 116740, 84002
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin)
0.19 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q8BGY6, Q99NB2, Q9BYG0
B3GNT5
B3GNT5 Fusion Protein Ag14246
20 μL
Primary
Rat, Mouse, Human
Antibody
IgG
Product Suggestions
Product Suggestions
Video's
SDS
Documentatie
Documentatie