missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AZI1 Rabbit anti-Human, Mouse, CoraLite 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Merk: Proteintech CL488-25735-100UL
This item is not returnable.
View return policy
Beschrijving
AZI1 Polyclonal antibody apecifically detetects AZI1 antigen in Human, Mouse Samples. It is validated for Flow Cytometry.Specificatie
AZI1 | |
Polyclonal | |
CoraLite 488 | |
BC136578 | |
Rabbit | |
Antigen affinity purification | |
Primary | |
Human, Mouse | |
Liquid |
Flow Cytometry | |
1000 μg/mL | |
PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3. | |
AZI1 | |
AZI1 fusion protein Ag22699 | |
100 μL | |
22994 | |
Store at -20°C. Avoid exposure to light. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction