Learn More
ASS1 Mouse anti-Human, CoraLite 488, Clone: 1D1A11, Proteintech
Mouse Monoclonal Antibody
Merk: Proteintech CL488-66036-100UL
This item is not returnable.
View return policy
Beschrijving
The ASS1 antibody from Proteintech is a mouse monoclonal antibody generated with fusion protein of human ASS1. This antibody recognizes human, pig, mouse, rat antigen. The ASS1 antibody has been validated for the following applications: IF analysis. This antibody is coralite™488.
Specificatie
ASS1 | |
Monoclonal | |
CoraLite 488 | |
100 μL | |
445 | |
IgG1 |
Immunofluorescence | |
1D1A11 | |
Mouse | |
Primary | |
Human, Mouse, Rat, Pig |