Learn More
ASS1 Mouse anti-Human, CoraLite 488, Clone: 1D1A11, Proteintech
Mouse Monoclonal Antibody
Merk:  Proteintech CL488-66036-100UL
Produktcode. 16860696
  • 450.00 /
Explore more special offers
Beschrijving
Beschrijving
The ASS1 antibody from Proteintech is a mouse monoclonal antibody generated with fusion protein of human ASS1. This antibody recognizes human, pig, mouse, rat antigen. The ASS1 antibody has been validated for the following applications: IF analysis. This antibody is coralite™488.
Specificatie
Specificatie
ASS1
Monoclonal
CoraLite 488
100 μL
445
IgG1
Immunofluorescence
1D1A11
Mouse
Primary
Human, Mouse, Rat, Pig
Product Suggestions
Product Suggestions
Video's
SDS
Documentatie
Documentatie
Certificates
Speciale aanbiedingen
Speciale aanbiedingen