Learn More
ANKZF1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Merk:  Proteintech 20447-1-AP-20UL
 Bekijk meerdere types van dit product via de
Produktcode. 16819994
  • 145.00 /
Beschrijving
Beschrijving
ZFR is critical for Staufen 2 isoform specific nucleocytoplasmic shuttling in neurons.
Specificatie
Specificatie
ANKZF1
Polyclonal
Unconjugated
ANKZF1
ANKZF1, Zinc finger protein 744, ZNF744
Rabbit
Antigen Affinity Chromatography
RUO
55139
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin)
0.51 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9H8Y5
ANKZF1
ANKZF1 Fusion Protein Ag14285