Learn More
ANKZF1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Merk: Proteintech 20447-1-AP-150UL
This item is not returnable.
View return policy
Beschrijving
ZFR is critical for Staufen 2 isoform specific nucleocytoplasmic shuttling in neurons.Specificatie
ANKZF1 | |
Polyclonal | |
Unconjugated | |
ANKZF1 | |
ANKZF1, Zinc finger protein 744, ZNF744 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
55139 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.51 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9H8Y5 | |
ANKZF1 | |
ANKZF1 Fusion Protein Ag14285 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |