Learn More
Sigma Laborzentrifugen™ Hoekrotors voor centrifuges uit de Sigma™ 2-serie
Used for sedimentation with high centrifugal forces typically needed in research laboratories. Sigma Laborzentrifugen™ Fixed Angle Rotors for Sigma™ Series 2 Centrifuges are precision-made from modern, certified high-performance materials with traceable batch numbers.
Specificatie
Type Angle Rotor
Meer specificaties weergeven
Beschrijving
Beschrijving
  • Can be continuously operated at maximum speed and +4°C in nearly all Sigma centrifuges
  • Can be used for very many cycles with maximum load and at maximum speed, in most cases up to 50,000 cycles
  • For use with a variety of bucket and tube sizes
Specificatie
Specificatie
Angle Rotor