missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aluminum Oxide, Activated Brockmann I, Honeywell Fluka™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
100 g
1 kg
5 kg
Packaging:
Poly Bottle
Unit Size:
100g
1kg
5kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Specifications
Specifications
Boiling Point | 2980°C |
Melting Point | 2072°C |
pH | Acidic |
Quantity | 1 kg |
Linear Formula | Al2O3 |
UN Number | NONH for all modes of transport |
IUPAC Name | dialuminum;oxygen(2-) |
Formula Weight | 101.96g/mol |
Packaging | Poly Bottle |
Appearance | Powder |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction