Learn More
Ansell™ AlphaTec™ 58-530B Series Nitrile Immersion Gloves
Enables users to handle wet or oily objects with less grip force and more control. The patented Ansell Grip TechnologyTM ensures that the coating polymer does not penetrate the liner during manufacture significantly reducing the likelihood of chemical leakage onto the skin.
Merk:  Ansell™ 58-530B_8
 Bekijk meerdere types van dit product via de
Produktcode. 15272529
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Bekijk hier alternatieve producten »
Beschrijving
Beschrijving
  • Nitrile polymer coating reduces chemical leakage onto skin
  • For indoor applications
  • Nylon liner
  • Gauntlet cuff
  • Sea green shell, anthracite grey dip color
TRUSTED_SUSTAINABILITY
Specificatie
Specificatie
Nitrile
Green, Gray
8
Gauntlet
Black Acrylic
Chemical resistance
Nitrile
Immersion Gloves
58-530B
Acrylic
6 Pairs
58-530B