Learn More
Alpha Tubulin Mouse anti-Human, Biotin, Clone: 1E4C11, Proteintech
Mouse Monoclonal Antibody
Merk: Proteintech BIOTIN-66031-100UL
This item is not returnable.
View return policy
Beschrijving
The Alpha Tubulin antibody from Proteintech is a mouse monoclonal antibody generated with fusion protein of human Alpha Tubulin. This antibody recognizes human, mouse, rat, canine antigen. The Alpha Tubulin antibody has been validated for the following applications: IHC analysis. This antibody is biotin.
Specificatie
Alpha Tubulin | |
Monoclonal | |
2000 μg/mL | |
Mouse | |
Primary | |
Human, Canine, Mouse, Rat |
Western Blot, Immunohistochemistry | |
1E4C11 | |
Biotin | |
100 μL | |
10376 | |
IgG2b |