Learn More
Schneider Ludwig™ Alcoholmeter
Merk: Schneider Ludwig™ 3312073
This item is not returnable.
View return policy
Beschrijving
Informatie over bestelling
dimLengthMetric
Specificatie
1Each | |
Alcohol Hydrometer | |
260 mm | |
0 to 100% |