Learn More
AGXT Mouse anti-Human, Unconjugated, Clone: 3E12H1, Proteintech
Mouse Monoclonal Antibody
Merk: Proteintech 67402-1-IG-150UL
This item is not returnable.
View return policy
Beschrijving
The AGXT antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human AGXT. This antibody recognizes human antigen. The AGXT antibody has been validated for the following applications: WB, ELISA analysis.
Specificatie
AGXT | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human |
Western Blot, ELISA | |
3E12H1 | |
Unconjugated | |
150 μL | |
189 | |
IgG1 |