Learn More
5-(tert-Butylcarbamoyl)pyridine-3-boronic acid, 98%, Thermo Scientific™
139.00 - 419.00
Chemische identificatiekenmerken
CAS | 913835-99-1 |
---|---|
Moleculaire formule | C10H15BN2O3 |
Molecular Weight (g/mol) | 222.051 |
MDL-nummer | MFCD08689533 |
InChI Key | FOJHBAIYLMKAIB-UHFFFAOYSA-N |
Synoniem | 5-tert-butylcarbamoyl pyridin-3-yl boronic acid, 5-tert-butylcarbamoyl pyridine-3-boronic acid, 3-t-butyl 5-borononicotinamide, 5-borono-n-tert-butyl nicotinamide, 5-tert-butylcarbamoyl pyridin-3-ylboronic acid, acmc-209rak, 3-tert-butyl 5-borononicotinamide, 5-t-butylcarbamoyl-3-pyridineboronic acid, 5-t-butylcarbamoyl pyridine-3-boronic acid |
PubChem CID | 44119356 |
IUPAC Name | [5-(tert-butylcarbamoyl)pyridin-3-yl]boronic acid |
SMILES | B(C1=CC(=CN=C1)C(=O)NC(C)(C)C)(O)O |
Beschrijving
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemische identificatiekenmerken
913835-99-1 | |
222.051 | |
FOJHBAIYLMKAIB-UHFFFAOYSA-N | |
44119356 | |
B(C1=CC(=CN=C1)C(=O)NC(C)(C)C)(O)O |
C10H15BN2O3 | |
MFCD08689533 | |
5-tert-butylcarbamoyl pyridin-3-yl boronic acid, 5-tert-butylcarbamoyl pyridine-3-boronic acid, 3-t-butyl 5-borononicotinamide, 5-borono-n-tert-butyl nicotinamide, 5-tert-butylcarbamoyl pyridin-3-ylboronic acid, acmc-209rak, 3-tert-butyl 5-borononicotinamide, 5-t-butylcarbamoyl-3-pyridineboronic acid, 5-t-butylcarbamoyl pyridine-3-boronic acid | |
[5-(tert-butylcarbamoyl)pyridin-3-yl]boronic acid |
Specificatie
5-(tert-Butylcarbamoyl)pyridine-3-boronic acid | |
245°C to 250°C | |
MFCD08689533 | |
FOJHBAIYLMKAIB-UHFFFAOYSA-N | |
[5-(tert-butylcarbamoyl)pyridin-3-yl]boronic acid | |
44119356 | |
98% |
913835-99-1 | |
C10H15BN2O3 | |
5-tert-butylcarbamoyl pyridin-3-yl boronic acid, 5-tert-butylcarbamoyl pyridine-3-boronic acid, 3-t-butyl 5-borononicotinamide, 5-borono-n-tert-butyl nicotinamide, 5-tert-butylcarbamoyl pyridin-3-ylboronic acid, acmc-209rak, 3-tert-butyl 5-borononicotinamide, 5-t-butylcarbamoyl-3-pyridineboronic acid, 5-t-butylcarbamoyl pyridine-3-boronic acid | |
B(C1=CC(=CN=C1)C(=O)NC(C)(C)C)(O)O | |
222.051 | |
222.05 |
Veiligheid en behandeling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
Your input is important to us. Please complete this form to provide feedback related to the content on this product.