Learn More
4-n-Hexyloxybenzeneboronic acid, 96%, Thermo Scientific™
52.00 - 201.00
Chemische identificatiekenmerken
CAS | 121219-08-7 |
---|---|
Moleculaire formule | C12H19BO3 |
Molecular Weight (g/mol) | 222.091 |
MDL-nummer | MFCD04039027 |
InChI Key | XYNVLFGOOWUKQS-UHFFFAOYSA-N |
Synoniem | 4-hexyloxyphenylboronic acid, 4-hexoxyphenyl boronic acid, 4-n-hexyloxy benzeneboronic acid, 4-hexyloxybenzeneboronic acid, 4-hexyloxy phenylboronic acid, 4-hexyloxy phenyl boronic acid, boronic acid,b-4-hexyloxy phenyl, 4-n-hexyloxy phenylboronic acid, pubchem9552 |
PubChem CID | 3682148 |
IUPAC Name | (4-hexoxyphenyl)boronic acid |
SMILES | B(C1=CC=C(C=C1)OCCCCCC)(O)O |
Beschrijving
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemische identificatiekenmerken
121219-08-7 | |
222.091 | |
XYNVLFGOOWUKQS-UHFFFAOYSA-N | |
3682148 | |
B(C1=CC=C(C=C1)OCCCCCC)(O)O |
C12H19BO3 | |
MFCD04039027 | |
4-hexyloxyphenylboronic acid, 4-hexoxyphenyl boronic acid, 4-n-hexyloxy benzeneboronic acid, 4-hexyloxybenzeneboronic acid, 4-hexyloxy phenylboronic acid, 4-hexyloxy phenyl boronic acid, boronic acid,b-4-hexyloxy phenyl, 4-n-hexyloxy phenylboronic acid, pubchem9552 | |
(4-hexoxyphenyl)boronic acid |
Specificatie
4-n-Hexyloxybenzeneboronic acid | |
C12H19BO3 | |
4-hexyloxyphenylboronic acid, 4-hexoxyphenyl boronic acid, 4-n-hexyloxy benzeneboronic acid, 4-hexyloxybenzeneboronic acid, 4-hexyloxy phenylboronic acid, 4-hexyloxy phenyl boronic acid, boronic acid,b-4-hexyloxy phenyl, 4-n-hexyloxy phenylboronic acid, pubchem9552 | |
B(C1=CC=C(C=C1)OCCCCCC)(O)O | |
222.091 | |
222.09 |
121219-08-7 | |
MFCD04039027 | |
XYNVLFGOOWUKQS-UHFFFAOYSA-N | |
(4-hexoxyphenyl)boronic acid | |
3682148 | |
96% |
Veiligheid en behandeling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
Your input is important to us. Please complete this form to provide feedback related to the content on this product.