missing translation for 'onlineSavingsMsg'
Learn More

4-Ethoxy-3-fluorobenzeneboronic acid, 98%, Thermo Scientific™

25.43 - 337.00

Chemische identificatiekenmerken

CAS 279263-10-4
Moleculaire formule C8H10BFO3
Molecular Weight (g/mol) 183.97
MDL-nummer MFCD04115667
InChI Key ZONJMULNQBNBGU-UHFFFAOYSA-N
Synoniem 4-ethoxy-3-fluorophenyl boronic acid, 4-ethoxy-3-fluorobenzeneboronic acid, 4-ethoxy-3-fluorophenylboronicacid, 3-fluoro-4-ethoxyphenyl boronic acid, boronic acid, 4-ethoxy-3-fluorophenyl, 4-ethoxy-3-fluoro-phenyl boronic acid, acmc-1cp7m, 4-borono-2-fluorophenetole, 3-fluoro-4-ethoxyphenylboronic acid, 4-ethoxy-3-fluorophenyl boronicacid
PubChem CID 2782821
IUPAC Name (4-ethoxy-3-fluorophenyl)boronic acid
SMILES CCOC1=CC=C(C=C1F)B(O)O
Meer specificaties weergeven

Producten 3
Produktcode Merk Aantal Prijs Hoeveelheid en beschikbaarheid  
Produktcode Merk Aantal Prijs Hoeveelheid en beschikbaarheid  
15439207
Bekijk documenten
Thermo Scientific Alfa Aesar
H52465.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15449207
Bekijk documenten
Thermo Scientific Alfa Aesar
H52465.06
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15459207
Bekijk documenten
Thermo Scientific Alfa Aesar
H52465.14
25 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Beschrijving

Beschrijving

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specificatie

Chemische identificatiekenmerken

279263-10-4
183.97
ZONJMULNQBNBGU-UHFFFAOYSA-N
2782821
CCOC1=CC=C(C=C1F)B(O)O
C8H10BFO3
MFCD04115667
4-ethoxy-3-fluorophenyl boronic acid, 4-ethoxy-3-fluorobenzeneboronic acid, 4-ethoxy-3-fluorophenylboronicacid, 3-fluoro-4-ethoxyphenyl boronic acid, boronic acid, 4-ethoxy-3-fluorophenyl, 4-ethoxy-3-fluoro-phenyl boronic acid, acmc-1cp7m, 4-borono-2-fluorophenetole, 3-fluoro-4-ethoxyphenylboronic acid, 4-ethoxy-3-fluorophenyl boronicacid
(4-ethoxy-3-fluorophenyl)boronic acid

Specificatie

4-Ethoxy-3-fluorobenzeneboronic acid
108°C to 113°C
MFCD04115667
ZONJMULNQBNBGU-UHFFFAOYSA-N
(4-ethoxy-3-fluorophenyl)boronic acid
2782821
98%
279263-10-4
C8H10BFO3
4-ethoxy-3-fluorophenyl boronic acid, 4-ethoxy-3-fluorobenzeneboronic acid, 4-ethoxy-3-fluorophenylboronicacid, 3-fluoro-4-ethoxyphenyl boronic acid, boronic acid, 4-ethoxy-3-fluorophenyl, 4-ethoxy-3-fluoro-phenyl boronic acid, acmc-1cp7m, 4-borono-2-fluorophenetole, 3-fluoro-4-ethoxyphenylboronic acid, 4-ethoxy-3-fluorophenyl boronicacid
CCOC1=CC=C(C=C1F)B(O)O
183.97
183.97
Video's
Veiligheid en behandeling

Veiligheid en behandeling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documentatie

Documentatie

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.