Learn More
4-Benzyloxy-3-methylbenzeneboronic acid, 96%, Thermo Scientific Chemicals
CAS: 338454-30-1 | C14H15BO3 | 242.081 g/mol
127.00 - 504.00
Chemische identificatiekenmerken
CAS | 338454-30-1 |
---|---|
Moleculaire formule | C14H15BO3 |
Molecular Weight (g/mol) | 242.081 |
MDL-nummer | MFCD06409944 |
InChI Key | QPXFEWQLALNXEZ-UHFFFAOYSA-N |
Synoniem | 4-benzyloxy-3-methylphenylboronic acid, 4-benzyloxy-3-methylphenyl boronic acid, 4-benzyloxy-3-methylbenzeneboronic acid, acmc-1agrw, 4-benzyloxy-3-methylphenylboronicacid, 4-benzyloxy-3-methyl-phenylboron acid, 4-benzyloxy-3-methylphenyl boronicacid |
PubChem CID | 17750522 |
IUPAC Name | (3-methyl-4-phenylmethoxyphenyl)boronic acid |
SMILES | B(C1=CC(=C(C=C1)OCC2=CC=CC=C2)C)(O)O |
Beschrijving
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemische identificatiekenmerken
338454-30-1 | |
242.081 | |
QPXFEWQLALNXEZ-UHFFFAOYSA-N | |
17750522 | |
B(C1=CC(=C(C=C1)OCC2=CC=CC=C2)C)(O)O |
C14H15BO3 | |
MFCD06409944 | |
4-benzyloxy-3-methylphenylboronic acid, 4-benzyloxy-3-methylphenyl boronic acid, 4-benzyloxy-3-methylbenzeneboronic acid, acmc-1agrw, 4-benzyloxy-3-methylphenylboronicacid, 4-benzyloxy-3-methyl-phenylboron acid, 4-benzyloxy-3-methylphenyl boronicacid | |
(3-methyl-4-phenylmethoxyphenyl)boronic acid |
Specificatie
4-Benzyloxy-3-methylbenzeneboronic acid | |
102°C to 106°C | |
MFCD06409944 | |
QPXFEWQLALNXEZ-UHFFFAOYSA-N | |
(3-methyl-4-phenylmethoxyphenyl)boronic acid | |
17750522 | |
96% |
338454-30-1 | |
C14H15BO3 | |
4-benzyloxy-3-methylphenylboronic acid, 4-benzyloxy-3-methylphenyl boronic acid, 4-benzyloxy-3-methylbenzeneboronic acid, acmc-1agrw, 4-benzyloxy-3-methylphenylboronicacid, 4-benzyloxy-3-methyl-phenylboron acid, 4-benzyloxy-3-methylphenyl boronicacid | |
B(C1=CC(=C(C=C1)OCC2=CC=CC=C2)C)(O)O | |
242.081 | |
242.08 |
Veiligheid en behandeling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.