Learn More
4'-Aminoacetophenone, 99%, Thermo Scientific Chemicals
CAS: 99-92-3 | C8H9NO | 135.17 g/mol
28.75 - 549.00
Chemische identificatiekenmerken
CAS | 99-92-3 |
---|---|
Moleculaire formule | C8H9NO |
Molecular Weight (g/mol) | 135.17 |
MDL-nummer | MFCD00007896 |
InChI Key | GPRYKVSEZCQIHD-UHFFFAOYSA-N |
Synoniem | 4'-aminoacetophenone, 4-aminoacetophenone, 1-4-aminophenyl ethanone, p-aminoacetophenone, 4-acetylaniline, p-acetylaniline, ethanone, 1-4-aminophenyl, p-aminoacetylbenzene, 1-4-aminophenyl ethan-1-one, acetophenone, p-amino |
PubChem CID | 7468 |
IUPAC Name | 1-(4-aminophenyl)ethanone |
SMILES | CC(=O)C1=CC=C(N)C=C1 |
Produktcode | Merk | Aantal | Prijs | Hoeveelheid en beschikbaarheid | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Merk | Aantal | Prijs | Hoeveelheid en beschikbaarheid | |||||
11369429
|
Thermo Scientific Alfa Aesar
A13213.14 |
25 g |
28.75
25 gram |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11490723
|
Thermo Scientific Alfa Aesar
A13213.18 |
50 g |
44.35
50 gram |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11400733
|
Thermo Scientific Alfa Aesar
A13213.30 |
250 g |
159.00
250 gram |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11480723
|
Thermo Scientific Alfa Aesar
A13213.0B |
1000 g |
549.00
1000 gram |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beschrijving
4'-Aminoacetophenone, is used in pharmaceuticals, medicines and other organic products. It is also used as a reagent for the photometric determination of Ce. As pharmaceutical intemediate or in the determination of palladium and vitaminB1.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4′-Aminoacetophenone, is used in pharmaceuticals, medicines and other organic products. It is also used as a reagent for the photometric determination of Ce. As pharmaceutical intemediate or in the determination of palladium and vitaminB1.
Solubility
It is soluble in hot water, ethanol and ether.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. acids, Incompatible to Acid chlorides, Acid anhydrides, Chloroformates, Strong oxidizing agents, Strong reducing agents. Stable under ordinary conditions.
Chemische identificatiekenmerken
99-92-3 | |
135.17 | |
GPRYKVSEZCQIHD-UHFFFAOYSA-N | |
7468 | |
CC(=O)C1=CC=C(N)C=C1 |
C8H9NO | |
MFCD00007896 | |
4'-aminoacetophenone, 4-aminoacetophenone, 1-4-aminophenyl ethanone, p-aminoacetophenone, 4-acetylaniline, p-acetylaniline, ethanone, 1-4-aminophenyl, p-aminoacetylbenzene, 1-4-aminophenyl ethan-1-one, acetophenone, p-amino | |
1-(4-aminophenyl)ethanone |
Specificatie
4'-Aminoacetophenone | |
103°C to 108°C | |
C8H9NO | |
471493 | |
4'-aminoacetophenone, 4-aminoacetophenone, 1-4-aminophenyl ethanone, p-aminoacetophenone, 4-acetylaniline, p-acetylaniline, ethanone, 1-4-aminophenyl, p-aminoacetylbenzene, 1-4-aminophenyl ethan-1-one, acetophenone, p-amino | |
GPRYKVSEZCQIHD-UHFFFAOYSA-N | |
1-(4-aminophenyl)ethanone | |
7468 | |
99% |
99-92-3 | |
292°C to 294°C | |
MFCD00007896 | |
14,413 | |
It is soluble in hot water,ethanol and ether. | |
CC(=O)C1=CC=C(N)C=C1 | |
135.17 | |
135.17 |
Veiligheid en behandeling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H312-H315-H319-H332-H335
missing translation for 'einecsNumber' : 202-801-2
missing translation for 'rtecsNumber' : AM5500000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.