Learn More
Memmert™ 4-20Ma Current Loop Interface Temperature 0-260°C Of One Pt100 In Chamber, For Uf260Ts
Memmert™ Temperature Output for external temperature monitoring.
Merk: Memmert™ UF260TSV6
438.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Beschrijving
- 4 to 20 mA current loop interface
- Temperature of a Pt100 sensor positioned flexibly in chamber for external temperature monitoring
Specificatie
Models 30, 55, 75, 110, 160, 260, 450, 750, 1060 | |
Current Loop Interface | |
4 to 20 mA |