Learn More
3'-Fluoro-4'-methoxyacetophenone, 99%, Thermo Scientific Chemicals
17.29 - 97.90
Chemische identificatiekenmerken
CAS | 455-91-4 |
---|---|
Moleculaire formule | C9H9FO2 |
Molecular Weight (g/mol) | 168.167 |
MDL-nummer | MFCD00026219 |
InChI Key | LQASUDVYVOFKNK-UHFFFAOYSA-N |
Synoniem | 3'-fluoro-4'-methoxyacetophenone, 3-fluoro-4-methoxyacetophenone, 1-3-fluoro-4-methoxyphenyl ethanone, 1-3-fluoro-4-methoxyphenyl ethan-1-one, ethanone, 1-3-fluoro-4-methoxyphenyl, 3'-fluoro-4-methoxyacetophenone, acetophenone, 3'-fluoro-4'-methoxy, 1-3-fluoro-4-methoxy-phenyl-ethanone, 1-3-fluoro-4-methoxyphenyl-1-ethanone, 1-3-fluoro-4-methoxy-phenyl ethanone |
PubChem CID | 96828 |
IUPAC Name | 1-(3-fluoro-4-methoxyphenyl)ethanone |
SMILES | CC(=O)C1=CC(=C(C=C1)OC)F |
Produktcode | Merk | Aantal | Prijs | Hoeveelheid en beschikbaarheid | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Merk | Aantal | Prijs | Hoeveelheid en beschikbaarheid | |||||
11443208
|
Thermo Scientific Alfa Aesar
A18972.03 |
1 g | N/A | ||||||
11453208
|
Thermo Scientific Alfa Aesar
A18972.06 |
5 g |
29.30
5 gram |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11463208
|
Thermo Scientific Alfa Aesar
A18972.14 |
25 g |
97.90
25 gram |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beschrijving
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemische identificatiekenmerken
455-91-4 | |
168.167 | |
LQASUDVYVOFKNK-UHFFFAOYSA-N | |
96828 | |
CC(=O)C1=CC(=C(C=C1)OC)F |
C9H9FO2 | |
MFCD00026219 | |
3'-fluoro-4'-methoxyacetophenone, 3-fluoro-4-methoxyacetophenone, 1-3-fluoro-4-methoxyphenyl ethanone, 1-3-fluoro-4-methoxyphenyl ethan-1-one, ethanone, 1-3-fluoro-4-methoxyphenyl, 3'-fluoro-4-methoxyacetophenone, acetophenone, 3'-fluoro-4'-methoxy, 1-3-fluoro-4-methoxy-phenyl-ethanone, 1-3-fluoro-4-methoxyphenyl-1-ethanone, 1-3-fluoro-4-methoxy-phenyl ethanone | |
1-(3-fluoro-4-methoxyphenyl)ethanone |
Specificatie
3'-Fluoro-4'-methoxyacetophenone | |
90°C to 94°C | |
C9H9FO2 | |
2084062 | |
LQASUDVYVOFKNK-UHFFFAOYSA-N | |
1-(3-fluoro-4-methoxyphenyl)ethanone | |
96828 | |
99% |
455-91-4 | |
147°C to 148°C (20 mmHg) | |
MFCD00026219 | |
3'-fluoro-4'-methoxyacetophenone, 3-fluoro-4-methoxyacetophenone, 1-3-fluoro-4-methoxyphenyl ethanone, 1-3-fluoro-4-methoxyphenyl ethan-1-one, ethanone, 1-3-fluoro-4-methoxyphenyl, 3'-fluoro-4-methoxyacetophenone, acetophenone, 3'-fluoro-4'-methoxy, 1-3-fluoro-4-methoxy-phenyl-ethanone, 1-3-fluoro-4-methoxyphenyl-1-ethanone, 1-3-fluoro-4-methoxy-phenyl ethanone | |
CC(=O)C1=CC(=C(C=C1)OC)F | |
168.167 | |
168.17 |
Veiligheid en behandeling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 207-253-8
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures