Learn More
Ohaus™ 2 x Centrifuge Buckets, Frontier™ R-S6x5/4 Swing Out Rotor
Carrier buckets for 5 mL centrifuge tubes, suitable for R-S6x5/4 Swing Out Rotors.
Brand: Ohaus™ 30130895
53.20 CHF valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Swinging bucket holds specifically sized tubes for centrifugation
- Suitable for use in Frontier™ R-S6x5/4 Swing Out Rotors
Specifications
Buckets, For 30130880 x2 | |
R-S6x5/4 Rotor | |
1 |
5 mL | |
Buckets for swing out rotor (2 pcs.) |