Learn More
Lenz Laborglasintrumente 2-Neck Adapters
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Merk: Lenz Laborglasintrumente 05000111
21.76 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Beschrijving
- With two NS sockets and one NS cone
- Ascending angle: 105°
- Made of DURAN™ tubing
Specificatie
Duran Glass | |
Socket Vertical NS 14/23 |