Acetonitrile (CH3CN)
Acetonitrile, CHROMASOLV™, for HPLC, gradient grade, ≥99.9%, Honeywell Riedel-de Haën™
CAS: 75-05-8 Molecular Formula: C2H3N Molecular Weight (g/mol): 41.053 MDL Number: MFCD00001878 InChI Key: WEVYAHXRMPXWCK-UHFFFAOYSA-N Synonym: mecn, methylkyanid, cyanure de methyl, acetonitrile, methane, cyano, ethyl nitrile, methanecarbonitrile, ethanenitrile, cyanomethane, methyl cyanide PubChem CID: 6342 ChEBI: CHEBI:38472 IUPAC Name: acetonitrile SMILES: CC#N
Acetonitrile, for HPLC, Fisher Chemical™
CAS: 75-05-8 MDL Number: 1878 InChI Key: WEVYAHXRMPXWCK-UHFFFAOYSA-N PubChem CID: 6342 ChEBI: CHEBI:38472 IUPAC Name: acetonitrile SMILES: CC#N
Acetonitrile, Optima™ LC/MS Grade, Fisher Chemical™
CAS: 75-05-8 Molecular Weight (g/mol): 41.053 MDL Number: 1878 InChI Key: WEVYAHXRMPXWCK-UHFFFAOYSA-N Synonym: mecn, methylkyanid, cyanure de methyl, acetonitrile, methane, cyano, ethyl nitrile, methanecarbonitrile, ethanenitrile, cyanomethane, methyl cyanide PubChem CID: 6342 ChEBI: CHEBI:38472 IUPAC Name: acetonitrile SMILES: CC#N
Acetonitrile, CHROMASOLV™, ≥99.9%, for LC-MS, Honeywell Riedel-de Haën™
CAS: 75-05-8 Molecular Formula: C2H3N Molecular Weight (g/mol): 41.053 MDL Number: MFCD00001878 InChI Key: WEVYAHXRMPXWCK-UHFFFAOYSA-N Synonym: mecn, methylkyanid, cyanure de methyl, acetonitrile, methane, cyano, ethyl nitrile, methanecarbonitrile, ethanenitrile, cyanomethane, methyl cyanide PubChem CID: 6342 ChEBI: CHEBI:38472 IUPAC Name: acetonitrile SMILES: CC#N
Acetonitrile CHROMASOLV™ Plus, for HPLC, ≥99.9%, Honeywell Riedel-de Haën™
CAS: 75-05-8 Molecular Formula: C2H3N Molecular Weight (g/mol): 41.053 MDL Number: MFCD00001878 InChI Key: WEVYAHXRMPXWCK-UHFFFAOYSA-N Synonym: mecn, methylkyanid, cyanure de methyl, acetonitrile, methane, cyano, ethyl nitrile, methanecarbonitrile, ethanenitrile, cyanomethane, methyl cyanide PubChem CID: 6342 ChEBI: CHEBI:38472 IUPAC Name: acetonitrile SMILES: CC#N
Acetonitrile, for UHPLC Gradient Grade Analysis
CAS: 75-05-8 Molecular Formula: C2H3N Molecular Weight (g/mol): 41.053 MDL Number: 1878 InChI Key: WEVYAHXRMPXWCK-UHFFFAOYSA-N Synonym: mecn, methylkyanid, cyanure de methyl, acetonitrile, methane, cyano, ethyl nitrile, methanecarbonitrile, ethanenitrile, cyanomethane, methyl cyanide PubChem CID: 6342 ChEBI: CHEBI:38472 IUPAC Name: acetonitrile SMILES: CC#N
Acetonitrile, HPLC for Gradient Analysis, meets analytical specification of Ph.Eur, Fisher Chemical™
CAS: 75-05-8 Molecular Formula: C2H3N MDL Number: 1878 InChI Key: WEVYAHXRMPXWCK-UHFFFAOYSA-N PubChem CID: 6342 ChEBI: CHEBI:38472 IUPAC Name: acetonitrile SMILES: CC#N
Acetonitrile, 99+%, for spectroscopy, Thermo Scientific Chemicals
CAS: 75-05-8 Molecular Formula: C2H3N Molecular Weight (g/mol): 41.053 MDL Number: MFCD00001878 InChI Key: WEVYAHXRMPXWCK-UHFFFAOYSA-N Synonym: mecn, methylkyanid, cyanure de methyl, acetonitrile, methane, cyano, ethyl nitrile, methanecarbonitrile, ethanenitrile, cyanomethane, methyl cyanide PubChem CID: 6342 ChEBI: CHEBI:38472 IUPAC Name: acetonitrile SMILES: CC#N
Acetonitrile, Advanced HPLC Grade for HPLC Gradient Analysis, Fisher Chemical™
CAS: 75-05-8 Molecular Formula: C2H3N Molecular Weight (g/mol): 41.05 MDL Number: 1878 InChI Key: WEVYAHXRMPXWCK-UHFFFAOYSA-N IUPAC Name: acetonitrile SMILES: CC#N
Acetonitrile, Extra Pure, SLR, Fisher Chemical™
CAS: 75-05-8 Molecular Formula: C2H3N Molecular Weight (g/mol): 41.05 MDL Number: 1878 InChI Key: WEVYAHXRMPXWCK-UHFFFAOYSA-N IUPAC Name: acetonitrile SMILES: CC#N
Chromplete™ Acetonitrile, HPLC/GC, meets ACS/USP analytical requirements, Thermo Scientific™
CAS: 75-05-8 Molecular Formula: C2H3N Molecular Weight (g/mol): 41.05 InChI Key: WEVYAHXRMPXWCK-UHFFFAOYSA-N Synonym: mecn, methylkyanid, cyanure de methyl, acetonitrile, methane, cyano, ethyl nitrile, methanecarbonitrile, ethanenitrile, cyanomethane, methyl cyanide PubChem CID: 6342 ChEBI: CHEBI:38472 IUPAC Name: acetonitrile SMILES: CC#N
Acetonitrile with 0.1% Trifluoroacetic Acid (v/v), Optima™ LC/MS Grade, Thermo Scientific™
CAS: 75-05-8 Molecular Formula: C2H3N Molecular Weight (g/mol): 41.053 MDL Number: 4169 InChI Key: WEVYAHXRMPXWCK-UHFFFAOYSA-N Synonym: mecn, methylkyanid, cyanure de methyl, acetonitrile, methane, cyano, ethyl nitrile, methanecarbonitrile, ethanenitrile, cyanomethane, methyl cyanide PubChem CID: 6342 ChEBI: CHEBI:38472 IUPAC Name: acetonitrile SMILES: CC#N
Acetonitrile, Spectrophotometric Grade, 99.7+%, Thermo Scientific Chemicals
CAS: 75-05-8 Molecular Formula: C2H3N Molecular Weight (g/mol): 41.053 MDL Number: MFCD00001878 InChI Key: WEVYAHXRMPXWCK-UHFFFAOYSA-N Synonym: mecn, methylkyanid, cyanure de methyl, acetonitrile, methane, cyano, ethyl nitrile, methanecarbonitrile, ethanenitrile, cyanomethane, methyl cyanide PubChem CID: 6342 ChEBI: CHEBI:38472 IUPAC Name: acetonitrile SMILES: CC#N
Acetonitrile, 99.9%, for residue analysis, Thermo Scientific Chemicals
CAS: 75-05-8 Molecular Formula: C2H3N Molecular Weight (g/mol): 41.053 MDL Number: MFCD00001878 InChI Key: WEVYAHXRMPXWCK-UHFFFAOYSA-N Synonym: mecn, methylkyanid, cyanure de methyl, acetonitrile, methane, cyano, ethyl nitrile, methanecarbonitrile, ethanenitrile, cyanomethane, methyl cyanide PubChem CID: 6342 ChEBI: CHEBI:38472 IUPAC Name: acetonitrile SMILES: CC#N
Acetonitrile, Certified, for HPLC-RMN with Low Propionitrile Level
CAS: 75-05-8 Molecular Formula: C2H3N Molecular Weight (g/mol): 41.05 MDL Number: 1878 InChI Key: WEVYAHXRMPXWCK-UHFFFAOYSA-N IUPAC Name: acetonitrile SMILES: CC#N
Acetonitrile, HPLC Grade (Far UV), 99.8+%, Thermo Scientific Chemicals
CAS: 75-05-8 Molecular Formula: C2H3N Molecular Weight (g/mol): 41.053 MDL Number: MFCD00001878 InChI Key: WEVYAHXRMPXWCK-UHFFFAOYSA-N Synonym: mecn, methylkyanid, cyanure de methyl, acetonitrile, methane, cyano, ethyl nitrile, methanecarbonitrile, ethanenitrile, cyanomethane, methyl cyanide PubChem CID: 6342 ChEBI: CHEBI:38472 IUPAC Name: acetonitrile SMILES: CC#N
Acetonitrile, Certified AR for Analysis, Fisher Chemical™
CAS: 75-05-8 Molecular Formula: C2H3N Molecular Weight (g/mol): 41.05 MDL Number: 1878 InChI Key: WEVYAHXRMPXWCK-UHFFFAOYSA-N IUPAC Name: acetonitrile SMILES: CC#N
Acetonitrile, 99.9%, Extra Dry over Molecular Sieve, AcroSeal™, Thermo Scientific Chemicals
CAS: 75-05-8 Molecular Formula: C2H3N Molecular Weight (g/mol): 41.053 MDL Number: MFCD00001878 InChI Key: WEVYAHXRMPXWCK-UHFFFAOYSA-N Synonym: mecn, methylkyanid, cyanure de methyl, acetonitrile, methane, cyano, ethyl nitrile, methanecarbonitrile, ethanenitrile, cyanomethane, methyl cyanide PubChem CID: 6342 ChEBI: CHEBI:38472 IUPAC Name: acetonitrile SMILES: CC#N
Acetonitrile, HPLC Grade, 99.7+% min, Thermo Scientific Chemicals
CAS: 75-05-8 Molecular Formula: C2H3N Molecular Weight (g/mol): 41.053 MDL Number: MFCD00001878 InChI Key: WEVYAHXRMPXWCK-UHFFFAOYSA-N Synonym: mecn, methylkyanid, cyanure de methyl, acetonitrile, methane, cyano, ethyl nitrile, methanecarbonitrile, ethanenitrile, cyanomethane, methyl cyanide PubChem CID: 6342 ChEBI: CHEBI:38472 IUPAC Name: acetonitrile SMILES: CC#N
Acetonitrile 99.8+%, for Residue Analysis, Distol™, Fisher Chemical™
CAS: 75-05-8 Molecular Formula: C2H3N Molecular Weight (g/mol): 41.05 MDL Number: 1878 InChI Key: WEVYAHXRMPXWCK-UHFFFAOYSA-N IUPAC Name: acetonitrile SMILES: CC#N
Acetonitrile, UHPLC-MS, Thermo Scientific™
Unique signal-to-noise specification for the chemical industry where the solvent quality is linked directly to the sensitivity of the mass spectrometer.
Acetonitrile, 99.6%, ACS reagent, Thermo Scientific Chemicals
CAS: 75-05-8 Molecular Formula: C2H3N Molecular Weight (g/mol): 41.053 MDL Number: MFCD00001878 InChI Key: WEVYAHXRMPXWCK-UHFFFAOYSA-N Synonym: mecn, methylkyanid, cyanure de methyl, acetonitrile, methane, cyano, ethyl nitrile, methanecarbonitrile, ethanenitrile, cyanomethane, methyl cyanide PubChem CID: 6342 ChEBI: CHEBI:38472 IUPAC Name: acetonitrile SMILES: CC#N
Acetonitrile, ACS, 99.5+%, Thermo Scientific Chemicals
CAS: 75-05-8 Molecular Formula: C2H3N Molecular Weight (g/mol): 41.053 MDL Number: MFCD00001878 InChI Key: WEVYAHXRMPXWCK-UHFFFAOYSA-N Synonym: mecn, methylkyanid, cyanure de methyl, acetonitrile, methane, cyano, ethyl nitrile, methanecarbonitrile, ethanenitrile, cyanomethane, methyl cyanide PubChem CID: 6342 ChEBI: CHEBI:38472 IUPAC Name: acetonitrile SMILES: CC#N
Acetonitrile, 99.5%, for analysis, Thermo Scientific Chemicals
CAS: 75-05-8 Molecular Formula: C2H3N Molecular Weight (g/mol): 41.053 MDL Number: MFCD00001878 InChI Key: WEVYAHXRMPXWCK-UHFFFAOYSA-N Synonym: mecn, methylkyanid, cyanure de methyl, acetonitrile, methane, cyano, ethyl nitrile, methanecarbonitrile, ethanenitrile, cyanomethane, methyl cyanide PubChem CID: 6342 ChEBI: CHEBI:38472 IUPAC Name: acetonitrile SMILES: CC#N
Acetonitrile (CH3CN), CAS 75-05-8, also known as methyl cyanide or cyanomethane, is the simplest organic nitrile. Produced mainly as a byproduct of manufacturing acrylonitrile, acetonitrile is a colorless liquid with an ether-like odor.
Available in various grades, acetonitrile is commonly used in the laboratory:
- As a polar aprotic solvent for a variety of organic reactions
- For such applications as high-performance liquid chromatography (HPLC)
- For reverse-phase HPLC as an alternative to methanol
In industry, it is likewise used as a solvent for the manufacture of such products as pharmaceuticals, pesticides, and perfumes.
When heated, acetonitrile emanates highly toxic hydrogen cyanide fumes and produces flammable vapors in reaction with water, steam, or acids. Exposure to acetonitrile in liquid or vapor form may irritate the eyes, nose, and respiratory tract. Long-term exposure may cause central nervous system damage. Handlers should wear protective gloves, proper eye and face protection, and avoid breathing fumes. Acetonitrile should be kept away from open flames, hot surfaces, and sources of ignition and stored in a well-ventilated, cool, tightly closed container.