Kernmonsternemers
Cole-Parmer™ Soil Auger Kit with Carrying Case
Soil Auger Kit with Carrying Case - Obtain 3 in. diameter samples in nearly all soil conditions with this complete auger kit.
Bel-Art™ SP Scienceware™ Tapered Plug Sampler
Punctures packaging to retrieve samples. Bel-Art™ SP Scienceware™ Tapered Plug Sampler is designed for sample extraction from bags or blocks of cheese, grease, soap, clay or similar semi-solids.
Buerkle™ Drill Head for Carcass Sampling
Drill head for carcass sampling has a hexagonal socket for bit holders or chucks.
Buerkle™ V4A MicroSampler Head
Buerkle™ V4A MicroSampler Head are interchangeable with different tips and volumes.
Buerkle™ Pigment Lance
Pigment lance to sample pigments, pastes and agglomerating bulk goods.
Sampling Systems™ MaxiCorer
Possesses a long curved blade that is ideal for use with powders and granules as well as for semi-solid materials such as cheese and thick grease. Sampling Systems™ MaxiCorer can be used as a sampler or for dispensing small quantities of product.
Buerkle™ V4A MicroSampler
Buerkle™ V4A MicroSampler designed for the sampling of small quantities of powder.
Buerkle™ Drill Bit for Buerkle™ Mole
Buerkle™ Drill Bit for Buerkle™ Mole are specially designed for very specific soil properties.
Buerkle™ Sealing Sleeves for Buerkle™ Zone Samplers
Sealing sleeves made of PP, sterilisable, they seal individual chambers when filling, emptying or transporting. The sealing sleeves are easy to slide on and they automatically clip into position.
Buerkle™ Silo Drill
If bulk goods are transported over long distances (in trucks, railway goods wagons), separation may occur.Taking the sample just from the surface therefore leads to the wrong result.Representative samples from all layers of the silo can be taken with the SiloDrill.
Buerkle™ Tubus™ Sampler
Buerkle™ Tubus™ Sampler makes it possible to revome bulk loads if foodstuff.
Sampling Systems™ SteriWare™ Sack and Bag Samplers
Sterile SteriWare™ Sack and Bag Samplers are the ideal tools for fast quality control work. Simply push into the sack to take the sample of powders and granules. Made from high grade HDPE with a sharp blade.
Buerkle™ Stainless Steel Scratcher
The scratcher has particularly been designed for sampling cohesive, caked and not free-flowing substances.
Buerkle™ V4A Stainless Steel StickProof Sampler
Features a conical inlet on the tip that allows the collection of either very small or larger quantities. Buerkle™ V4A Stainless Steel StickProof Sampler constructed in one piece without edges, grooves or crevices for perfect and easy cleaning.
Buerkle™ Stainless Steel Core Sampler
Can be used to take samples of bulk materials which can easily become stuck or settle during sampling due to their particle form.
Sampling Systems™ MiniCorer
Suitable for sampling sticky, cohesive powders as well as products such as cheese. Sampling Systems™ MiniCorer is excellent at taking samples from semi solid products.
Buerkle™ Sampler PowderProof™
Sampler PowderProof™ is the practically proven professional solution for mobile sampling of powders and granules.
Buerkle™ ViscoSampler
ViscoSampler with powerful suction for viscous media. The media is drawn up and discharged using the suction flask. Specially developed for viscous substances such as slurries, silt, wet clay and soil samples, soil sediments, oils, emulsions, creams etc.
Buerkle™ Stainless Steel Ice Samplers
Sampler developed for sampling from frozen, highly viscous, paste-like materials such as ice-cream and similar substances. The drill-shaped cutter screws itself into the frozen goods independently when being rotated.The collection cylinder can easily be detached via a bayonet connection.
Buerkle™ Aluminum All-Layer-Sampler
Buerkle™ Aluminum All-Layer-Sampler allows samples to be collected from every layers of the container.
Buerkle™ Stainless Steel Quali Rod Cylindrical
Cylindrical hollow drill with a sharp cutting edge, can easily be driven into paste-like and semi-solid substances. The samples cut out are subsequently removed with the appropriate push-rod ejector. Total length 260 mm. Inner Ø 8 mm, outer Ø 10 mm.
Buerkle™ QualiSampler™ Set
The handy set, which comes complete with carrying case, combines the LiquiSampler and the ViscoSampler with practical accessories.Models in PP or PTFE/ FEP.
Buerkle™ QuickPicker
The core sampler QuickPicker is particularly suitable in the event that bulk goods are taken directly from sacks or bags and have to be filled into the original sampling bottle directly. QuickPicker is uncomplicated in its use and cleaning.
Buerkle™ Spiral Screw for PowderProof™ Sampler
PowderProof is the practically proven professional solution for mobile sampling of powders and granules. Different lances and augers can be attached to the robust drive unit and swapped without difficulty. Stainless steel V4A (1.4404)
Buerkle™ Uitschuifbare stang
Telescopic Rod is a versatile sampling system with exchangeable tools for a wide range of applications.
Buerkle™ Stainless Steel Ice Borer
Buerkle™ Stainless Steel Ice Borer is a sampler for deep-frozen and semi-solid substances products in the food industry.
Buerkle™ Silo Sampler SiloPicker
Buerkle™ Stainless Steel SiloPicker Sampler is ideal for taking bulk goods samples from silos.
Buerkle™ V4A Stainless Steel Mini ViscoSampler
Buerkle™ V4A Stainless Steel Mini ViscoSampler for viscous substances such as pastes, mud, creams, oils.
Buerkle™ GeoSampler Soil Sampler
Buerkle™ GeoSampler Soil Sampler is for light soils, meadows and lawns.
Buerkle™ V4A Stainless Steel PharmaPicker
The V4A Stainless Steel PharmaPicker is for sampling very small quantities of highly potent, expensive or active powders for analysis.