Learn More
Gosselin™ White Specimen Containers with Red Cap and Spatula
Corning™ Gosselin™ Straight Container, Specimen Vials, PP come in different sizes and quantities, and are available sterile and non-sterile.
Merk: Gosselin™ TP52C-022
This item is not returnable.
View return policy
Beschrijving
- Clear and break-resistant polypropylene (PP)
- Available in different sizes and quantities
- Color-coded screw caps
Specificatie
Specimen Container | |
Polypropylene | |
Non-sterile | |
Red | |
White | |
No Label | |
380/Pk. |
Yes | |
Straight | |
125 mL | |
Screw Cap | |
Cap with white spoon | |
380/Cs. |