Learn More
Aalborg™ Directeflowbuis, water
Aalborg™ Water Direct Glass Flow Tube completes a flowmeter, which is used in fluid systems to indicate the rate of flow of the fluid.
Merk: Aalborg™ FT-112-05-ST-VN
This item is not returnable.
View return policy
Beschrijving
- Direct reading tubes are straightforward.
- The measurement scale on each of these tubes reads actual flow at standard conditions in a choice of English or Metric units.
- Obtaining actual flow rates with these tubes requires the use of a reference scale flow correlation table.
- Reference scale tubes are useful when measuring flow rates for gases other than air and/or for non-standard conditions.
Specificatie
20 mL/min. | |
20mL/min. | |
Water Direct Flow Tube | |
150 mm |