Learn More
Thermo Scientific Freezer -86°C ultra-low temperature TSU300V 420.9L
Merk:  Thermo Scientific TSU300V
Freezer -86°C ultra-low temperature TSU300V 420.9L
Overige gegevens : Weight : 303.00000kg
Produktcode. 13557150
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Bekijk hier alternatieve producten »
Explore more special offers