Learn More
Bunzl Healthcare™ Terumo™-injectiespuit met excentrische Luer-Slip-aansluiting
Manufactured with polypropylene for superior thermal and chemical resistance. Bunzl™ Terumo™ Eccentric Luer-Slip Syringes have a unique bevel design to facilitate the penetration of the skin and minimize trauma.
Merk: Bunzl Healthcare™ SS20ES1
This item is not returnable.
View return policy
Beschrijving
- Clear polypropylene barrel and plunger; double contact ring
- Integrated plunger stop prevents accidental pull-out
- Sterile, Latex-free
- Bold, precise scale markings
- Complies to ISO 7886 (DIS 7886)
- Individually wrapped in tamper evident peel packs
- Supplied without needles
- Suitable for gases or liquids
- Eccentric luer lock
For research use only, unless otherwise indicated.
Specificatie
ISO 7886 (DIS 7886) | |
Voor onderzoeksdoeleinden, voor gassen en vloeistoffen | |
Ja | |
50 per pak | |
Excentrische Luer-Slip | |
Ja |
Polypropylene | |
Injectiespuit met excentrische Luer-Slip-aansluiting | |
1 ml | |
20 ml | |
Ja |