Learn More
Welch™ Synthetic Clamping Chain
Merk:  Welch™ 710653-1
Produktcode. 10210272
  • 40.70 / 1 stuk
Explore more special offers
Toevoegen aan winkelmandje
Beschrijving
Beschrijving
TRUSTED_SUSTAINABILITY
Specificatie
Specificatie
1 Ea.
Synthetic Clamping Chain
23 mm
Product Suggestions
Product Suggestions
Video's
SDS
Documentatie
Documentatie
One moment while we fetch your results.
Certificates
Speciale aanbiedingen
Speciale aanbiedingen
Product Content Correction
Product Title
Welch™ Synthetic Clamping Chain > Dimensions: 52 x 16 x 48mm