Learn More
Superfos™ SuperLift™ polypropyleen standaard met centreerdeksel
Manufactured from polypropylene. Superfos™ SuperLift™ Polypropylene Standard with Centering Lid includes tamper evidence while the protective collar safeguards the lid to prevent container ovalisation.
Merk: Superfos™ 4083
This item is not returnable.
View return policy
Beschrijving
- Manufactured from polypropylene
- Includes tamper evidence while the protective collar safeguards the lid and prevents container ovalisation.
- Very good heat resistance
- Packaging is suitable for hot filling.
- Notably, plastic packaging become flexible when subjected to hot filling temperatures and care must be taken in relation to stacking immediately after hot filling.
- Freezing conditions require special resins available on request.
Specificatie
Standaard met centreringsdeksel | |
Ja | |
265.8 mm | |
Standaard met centreringsstukken |
Polypropyleen | |
Gewijzigde Verordening nr. 10/2011/EG van de Europese Commissie | |
13.3 mm | |
Pakket 1 |