Learn More
Superfos™ SuperLift™ polypropyleen standaard met centreerdeksel
Manufactured from polypropylene. Superfos™ SuperLift™ Polypropylene Standard with Centering Lid includes tamper evidence while the protective collar safeguards the lid to prevent container ovalisation.
Merk:  Superfos™ 4083
Encompass
 Bekijk meerdere types van dit product via de
Produktcode. 10195044
  • 0.69 /
Explore more special offers
Toevoegen aan winkelmandje
Beschrijving
Beschrijving
  • Manufactured from polypropylene
  • Includes tamper evidence while the protective collar safeguards the lid and prevents container ovalisation.
  • Very good heat resistance
  • Packaging is suitable for hot filling.
  • Notably, plastic packaging become flexible when subjected to hot filling temperatures and care must be taken in relation to stacking immediately after hot filling.
  • Freezing conditions require special resins available on request.
Specificatie
Specificatie
Standaard met centreringsdeksel
Ja
265.8 mm
Standaard met centreringsstukken
Polypropyleen
Gewijzigde Verordening nr. 10/2011/EG van de Europese Commissie
13.3 mm
Pakket 1
Product Suggestions
Product Suggestions
Video's
SDS
Documentatie
Documentatie
Certificates
Speciale aanbiedingen
Speciale aanbiedingen