Learn More
IBA Lifesciences Strep-Tactin™ Superflow™, 50% Suspension
Used for purification of recombinant Strep-tag™ proteins under gravity flow and low pressure or FPLC
Merk:  IBA Lifesciences 2-1206-500
Produktcode. 12886446
Beschrijving
Beschrijving
  • Bulk resin is recommended for packing of columns and cartridges
  • Resin is optimized for column affinity chromatography rather than for batch purification
  • Binding capacity: 1 mL sedimented resin (corresponding to 2 mL of 50% suspension)
  • Useful for one-step purification of 50 to 100 nmol recombinant Strep-tag™ protein (up to 3 mg in case of 30 kDa protein)
Specificatie
Specificatie
Strep-Tactin Superflow
Product Suggestions
Product Suggestions
Video's
SDS
Documentatie
Documentatie
One moment while we fetch your results.
Certificates
Speciale aanbiedingen
Speciale aanbiedingen
Product Content Correction
Product Title
IBA Lifesciences Strep-Tactin™ Superflow™, 50% Suspension >