Learn More
Swann-Morton™ Stainless Steel Individually Wrapped, Non-Sterile Scalpel Handles
Longer version of the No. 3 handle.
Merk: Swann-Morton™ 913
This item is not returnable.
View return policy
Beschrijving
- Longer version of the No. 3 handle.
- Fits the following blades:
- No. 6, 9, 10, 10A, E/11, 11, 11P, 12, 12D, 13, 14, 15, 15A, 15C, 15T, 16, 40, SG3.
- Individually wrapped and boxed in units of 10.
Specificatie
Stainless Steel | |
Scalpel Handle | |
3 LS/S | |
Non-sterile |