Learn More
Socorex™ Stainless Steel Bent Drenchers
Merk: Socorex™ 376.1251B
32.40 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Specificatie
Autoclavable | |
Stainless Steel | |
51 mm |
Bent Drencher | |
For Safe Animal Nose and Mouth Feeding |