Learn More
SVL™ Rotulex™ Spherical Joints
Offers a grease free spherical joint
Merk: SVL™ RF19
This item is not returnable.
View return policy
Beschrijving
- All glass and PTFE pathway is highly resistant to chemical attack
- Can be used at temperatures up to 200°C
- System comprises male ball joint with PTFE covered O-ring which fits into female cup joint
- Joint can be retained together with spring loaded metal clip
- Component parts RM, RF, RO and RC should be ordered separately
Specificatie
Female joint | |
Glass, PTFE | |
Spherical joint | |
19 |
Product Content Correction
Product Title
SVL™ Rotulex™ Spherical Joints > Bore size: 9mm; Joint size: 19