Learn More
Buerkle™ Polyethylene Flat Seal
Flat seals made of Polyethylene, transparent fitting outer or inner thread.
Merk: Buerkle™ 8538-0100
7.38 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Beschrijving
- PE
Specificatie
Transparent | |
1 in. thread | |
Polyethylene | |
Seal, Flat |