Learn More
Fisherbrand™ Vlakke PTFE-roerstaven
Provides stable stirring action. Fisherbrand™ Plain PTFE Stir Bars provide greater turbulence than cylindrical stir bars.
Merk: Fisherbrand™ 001.240
This item is not returnable.
View return policy
Beschrijving
- Stable, general-purpose magnetic stir bar
- Provides greater turbulence than cylindrical stir bars
- Autoclavable