Learn More
SI Analytics™ pH Combination Electrode
Ensure high measurement accuracy, stability, and long service life while maintaining high adaptability to your measurement tasks. SI Analytics™ pH Combination Electrode is robust, durable, and easy to clean.
80.75 - 523.00
Specificatie
pH-bereik | 0 to 14 |
---|---|
Membraan | Sphere |
Diameter (metrisch) | 12 mm |
Produktcode | Merk | Lengte (metrisch) | Knooppunt | Temperatuurbereik (metrisch) | Materiaal | Prijs | Hoeveelheid en beschikbaarheid | ||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Merk | Lengte (metrisch) | Knooppunt | Temperatuurbereik (metrisch) | Materiaal | Prijs | Hoeveelheid en beschikbaarheid | ||
10558281
|
SI Analytics™
285100042 |
120 mm | Keramisch membraan | -5°C to +100°C | A-glas |
282.00
1 stuk |
|
||
10342911
|
SI Analytics™
285100034 |
120 mm | Platinamembraan | -5°C to +100°C | A-glas |
297.00
1 stuk |
|
||
10131721
|
SI Analytics™
285102524 |
170 mm | Keramisch membraan | 0°C to 100°C | H-glas |
302.00
1 stuk |
|
||
10383001
|
SI Analytics™
285100018 |
170 mm | Keramisch membraan | -5°C to +100°C | A-glas |
289.00
Each |
|
||
10040171
|
SI Analytics™
285100231 |
170 mm | Slijpstukmembraan | 0°C to 100°C | H-glas |
368.00
1 stuk |
|
||
10622832
|
SI Analytics™
285092329 |
170 mm | Slijpstukmembraan | 0°C to 40°C | A-glas |
372.00
1 stuk |
|
||
10131831
|
SI Analytics™
285092337 |
170 mm | Slijpstukmembraan | 10°C to 40°C | A-glas |
372.00
1 stuk |
|
||
10434261
|
SI Analytics™
285100059 |
170 mm | Slijpstukmembraan | –5 tot 100 °C | A-glas |
450.00
Each |
|
||
10057790
|
SI Analytics™
285100001 |
170 mm | Platinamembraan | –5 tot 100 °C | A-glas |
374.00
1 stuk |
|
||
10189750
|
SI Analytics™
285100207 |
170 mm | Platinamembraan | 0°C to 100°C | H-glas |
307.00
1 stuk |
|
||
10408992
|
SI Analytics™
285102549 |
320 mm | Keramisch membraan | 0°C to 100°C | H-glas |
336.00
1 stuk |
|
||
10403891
|
SI Analytics™
285100223 |
320 mm | Platinamembraan | 0 tot 100 °C | H-glas |
339.00
1 stuk |
|
||
10092812
|
SI Analytics™
285092591 |
600 mm | Platinamembraan | 0°C to 100°C | H-glas |
523.00
1 stuk |
|
||
10492452
|
SI Analytics™
285092583 |
500 mm | Platinamembraan | 0 tot 100 °C | H-glas |
490.00
1 stuk |
|
||
10129052
|
SI Analytics™
285130220 |
170 mm | Ground Joint Diaphragm | 0°C to 100°C | H-Glass | ||||
Beschrijving
- Proven high-end electrodes for demanding measurement tasks
- Double junction Silamid™ reference system for fast and stable acquiring of measured values and for longer electrode life
- Utmost versatility of pH electrodes is achieved by a large selection of junctions, membrane glass types and shapes, shaft lengths and diameters, ground joints, plug connections and integrated temperature sensors
- Each pH combination electrode comes with individual serial number and quality certificate
Specificatie
0 to 14 | |
Sphere | |
12 mm |