Learn More
Liebherr™ LGv 5010 MediLine Freezer
Protects valuable contents with a variety of alarm, memory and safety features. Liebherr™ LGv 5010 MediLine Freezer features the Comfort electronic controller, advanced cooling technology, multiple organizing drawers/bins, and an easy-clean molded plastic interior.
Merk: Liebherr™ LGV 5010
This item is not returnable.
View return policy
Beschrijving
- Integrated Alarms:
- Audio and visual temperature, door opening and power failure alarms, as well internal system alarms (e.g. probe)
- Alarm Test Function: Check whether an internally or externally connected alarm system is functioning correctly without impacting refrigeration performance
- Comfort Controller with Digital Display: Internal memory stores details of last three alarms and high/low temperatures
- Serial Bus Interface: Electronic control system uses an RS 485 interface to connect up to 20 appliances to external monitoring software
- Temperature Stability EN 60068-3: Measures the temperature at nine points in the interior according to procedure EN 60068-3, using state-of-the-art facilities
- Forced Air Cooling: Highly efficient fans quickly achieves and maintains an even temperature throughout the interior
- Efficient Refrigeration System: Reduces energy consumption and operating costs with premium-quality components
- Environmentally Friendly Refrigerant: Minimal greenhouse effect and no ozone depletion potential with outstanding energy efficiency