Learn More
Labconco™ Support Shelves: Telescoping Base Stand
For use with Telescoping Base Stands supporting Purifier Logic Biosafety Cabinets and PuriCare Procedure Stations.
Beschrijving
Beschrijving
For use with Telescoping Base Stands supporting Purifier Logic Biosafety Cabinets and PuriCare Procedure Stations (pre-2014). Support Shelf provides 8.0 in. x 20.0 in. (20 x 51 cm) work surface to support remote equipment such as an aspirator pump within easy access of the biosafety cabinet. Powder-coated steel shelf mounts to left-side tubular frame of the base stand. Includes powder-coated steel shelf and fasteners. Not for use with SoLo Hydraulic Lift Base Stand. Installation is required.